Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amantadine ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A136871-100G A136871-1G A136871-25G A136871-5G
ALA136871-100GEA 403.66 USD InStock
ALA136871-100G ALA136871-1G ALA136871-25G ALA136871-5G
Amantadine ≥98%, Moligand™
Amantadine
Building block for an L-piperidinamide catalyst used in an enantioselective Strecker reaction of phosphinoyl imines.application:1-Adamantanamine acts as a catalyst in enantioselective strecker reaction of phosphinoyl imines. It is also useful for making adamantane derivatives, i.e. amantadine hydrochloride. It is used in the pharmaceutical industry as antiviral and an antiparkinsonian drug.


Formula: C₁₀H₁₇N
MW: 151.25 g/mol
Boiling Pt: 214
Melting Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00074732
CAS Number: 768-94-5
EINECS: 212-201-2
Merck Index: 12,00389
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty