Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

β-Alanine 98, Moligand™

Supplier: ALADDIN SCIENTIFIC

A137901-250G A137901-25G A137901-100G
ALA137901-250GEA 21.76 USD InStock
ALA137901-250G ALA137901-25G ALA137901-100G
β-Alanine 98, Moligand™
β-Alanine
β-Alanine has been used as a ligand for the orphan MAS-related receptor.[1] It has also been used in culture media used for certain strains of yeast to test for β-alanine auxotrophy.


Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201
Melting Pt: 94
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty