Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apigetrin, cosmosiin ≥98%

Supplier: ALADDIN SCIENTIFIC

A139025-25MG A139025-5MG
ALA139025-25MGEA 327.66 USD InStock
ALA139025-25MG ALA139025-5MG
Apigetrin, cosmosiin ≥98%
Apigetrin, cosmosiin
Application:Apigenin 7-Glucoside is a by-product during the synthesis of Isorhoifolin (I819700), a naturally occurring flavonoid that was shown to exhibit potential antidiabetic, antihyperlipidemic and antioxidant effects.


Formula: C₂₁H₂₀O₁₀
Storage Temperature: Ambient
MDL Number: MFCD00016787
CAS Number: 578-74-5
EINECS: 209-430-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty