Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminobutyraldehyde diethyl acetal ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A139122-100G A139122-1G A139122-25G A139122-5G
ALA139122-100GEA 555.66 USD InStock
ALA139122-100G ALA139122-1G ALA139122-25G ALA139122-5G
4-Aminobutyraldehyde diethyl acetal ≥97% (by GC)
4-Aminobutyraldehyde diethyl acetal


Formula: C₈H₁₉NO₂
MW: 161.24 g/mol
Boiling Pt: 196 °C
Flash Pt: 62°C
Storage Temperature: Ambient
MDL Number: MFCD00008227
CAS Number: 6346-09-4
EINECS: 228-753-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty