Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(trifluoromethyl)benzonitrile ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A140249-100G A140249-1G A140249-25G A140249-5G
ALA140249-100GEA 2222.91 USD InStock
ALA140249-100G ALA140249-1G ALA140249-25G ALA140249-5G
2-Amino-4-(trifluoromethyl)benzonitrile ≥98% (by GC)
2-Amino-4-(trifluoromethyl)benzonitrile


Formula: C₈H₅F₃N₂
MW: 186.1339 g/mol
Boiling Pt: 127°C
Melting Pt: 88.0-92.0°C
MDL Number: MFCD04974124
CAS Number: 1483-54-1
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty