Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Aminomethyl)phosphonic acid ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

A151006-1G A151006-250MG A151006-25G A151006-5G A151006-100G
ALA151006-1GEA 55.96 USD InStock
ALA151006-1G ALA151006250MG ALA151006-25G ALA151006-5G ALA151006-100G
(Aminomethyl)phosphonic acid ≥97% (by titration)
(Aminomethyl)phosphonic acid
(Aminomethyl)phosphonic acid is a metabolite of amitrole and glyphosate herbicides and was analyzed by HPLC using an electrochemical detector.(Aminomethyl)phosphonic acid was used to develop a novel kind of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.


Formula: CH₆NO₃P
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS Number: 1066-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty