Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-phenyl-1,3,4-thiadiazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151014-1G A151014-250MG A151014-5G
ALA151014-1GEA 73.06 USD InStock
ALA151014-1G ALA151014250MG ALA151014-5G
2-Amino-5-phenyl-1,3,4-thiadiazole ≥98%
2-Amino-5-phenyl-1,3,4-thiadiazole
2-Amino-5-phenyl-1,3,4-thiadiazole on condensation with benzaldehyde (SPT), 4-nitrobenzaldehyde (SNT), 4-methoxybenzaldehyde (SMT), 2-hydroxybenzaldehyde (SSTH) or 2-hydroxyacetophenone affords Schiff′s bases. Its molecular geometry and vibrational frequencies have been evaluated using the Hartree-Fock and density functional method (B3LYP). 2-Amino-5-phenyl-1,3,4-thiadiazole has been reported to inhibit the corrosion of mild steel in 0.5M H2SO4 and 1.0M HCl. Corrosion inhibition has been examined using potentiodynamic polarization and electrochemical impedance spectroscopy (EIS).


Formula: C₈H₇N₃S
Melting Pt: 223-230 °C
Storage Temperature: Ambient
MDL Number: MFCD00205278
CAS Number: 2002-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty