Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone ≥98%

Supplier: ALADDIN SCIENTIFIC

A151023-1G A151023-250MG A151023-25G A151023-5G
ALA151023-1GEA 79.71 USD InStock
ALA151023-1G ALA151023250MG ALA151023-25G ALA151023-5G
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone ≥98%
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone


Formula: C₁₁H₁₃N₃O
Melting Pt: 196 °C
MDL Number: MFCD01692712
CAS Number: 36725-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty