Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetamidobenzaldehyde ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151026-1G A151026-250MG A151026-5G
ALA151026-1GEA 521.46 USD InStock
ALA151026-1G ALA151026250MG ALA151026-5G
2-Acetamidobenzaldehyde ≥95%
2-Acetamidobenzaldehyde


Formula: C₉H₉NO₂
Boiling Pt: 103°C
Melting Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD06739581
CAS Number: 13493-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty