Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151033-1G A151033-250MG A151033-5G
ALA151033-1GEA 75.91 USD InStock
ALA151033-1G ALA151033250MG ALA151033-5G
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine ≥97%
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine


Formula: C₇H₆N₄O
Melting Pt: 116-118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075383
CAS Number: 107866-54-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty