Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline-5-amine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151038-100G A151038-1G A151038-25G A151038-5G
ALA151038-100GEA 790.31 USD InStock
ALA151038-100G ALA151038-1G ALA151038-25G ALA151038-5G
Isoquinoline-5-amine ≥98% (by GC)
Isoquinoline-5-amine
5-Aminoisoquinoline forms 1:1 host?guest inclusion complex with β-cyclodextrin. It enhances the chemiluminescence of luminol-H2O2-horseradish peroxidase.5-Aminoisoquinoline (5AIQ) was used to study the effect of addition of β-cyclodextrin on the absorption and emission properties of 5AIQ


Formula: C₉H₈N₂
Melting Pt: 125-128 °C
Storage Temperature: Ambient
MDL Number: MFCD00006907
CAS Number: 1125-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty