Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl ethyl ether ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151091-1ML A151091-25ML A151091-5ML
ALA151091-1MLEA 12.26 USD InStock
ALA151091-1ML ALA151091-25ML ALA151091-5ML
Allyl ethyl ether ≥95% (by GC)
Allyl ethyl ether
Application:Allyl ethyl ether is used as an intermediate to manufacture other chemicals.


Formula: C₅H₁₀O
Boiling Pt: 67°C
Density: 0.77 g/cm³
Flash Pt: -20°C
Storage Temperature: Refrigerator
MDL Number: MFCD00009255
CAS Number: 557-31-3
UN: 2335
ADR: 3, 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty