Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl phenylacetate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151106-1ML A151106-25ML A151106-5ML
ALA151106-1MLEA 9.41 USD InStock
ALA151106-1ML ALA151106-25ML ALA151106-5ML
Allyl phenylacetate ≥98% (by GC)
Allyl phenylacetate


Formula: C₁₁H₁₂O₂
Boiling Pt: 240°C
MDL Number: MFCD00048176
CAS Number: 1797-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty