Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-6-aminopyridin-2-ylcarbamate ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Warning

A151149-5G A151149-1G A151149-25G
ALA151149-5GEA 328.61 USD InStock
ALA151149-5G ALA151149-1G ALA151149-25G
tert-Butyl-6-aminopyridin-2-ylcarbamate ≥97% (by titration)
tert-Butyl-6-aminopyridin-2-ylcarbamate


Formula: C₁₀H₁₅N₃O₂
MW: 209.2500 g/mol
Melting Pt: 127 °C
MDL Number: MFCD09037865
CAS Number: 322690-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty