Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetoxypropionic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151194-1G A151194-250MG A151194-25G A151194-5G
ALA151194-1GEA 74.01 USD InStock
ALA151194-1G ALA151194250MG ALA151194-25G ALA151194-5G
2-Acetoxypropionic acid ≥98%
2-Acetoxypropionic acid


Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 127°C
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674491
CAS Number: 535-17-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty