Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetyl-1-methyl-1H-pyrrole ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

A151233-25G A151233-5G
ALA151233-25GEA 124.36 USD InStock
ALA151233-25G ALA151233-5G
2-Acetyl-1-methyl-1H-pyrrole ≥98% (by GC)
2-Acetyl-1-methyl-1H-pyrrole
2-Acetyl-1-methylpyrrole was employed as model pyrrole to investigate the antioxidative activity of nonenzymatic browning reactions.[1] It was employed as catalyst for the sulfonylation of spectinomycin


Formula: C₇H₉NO
Boiling Pt: 201℃
Density: 1.04 g/cm³
Flash Pt: 82°C
MDL Number: MFCD00003089
CAS Number: 932-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty