Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-chloropyrimidin-4(3H)-one ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151266-1G A151266-250MG A151266-25G A151266-5G
ALA151266-1GEA 22.71 USD InStock
ALA151266-1G ALA151266250MG ALA151266-25G ALA151266-5G
2-Amino-6-chloropyrimidin-4(3H)-one ≥98% (by HPLC)
2-Amino-6-chloropyrimidin-4(3H)-one


Formula: C₄H₄ClN₃O
MW: 145.5500 g/mol
MDL Number: MFCD00006093
CAS Number: 1194-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty