Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetoxycinnamic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151300-1G A151300-250MG A151300-5G
ALA151300-1GEA 23.66 USD InStock
ALA151300-1G ALA151300250MG ALA151300-5G
2-Acetoxycinnamic acid ≥95%
2-Acetoxycinnamic acid


Formula: C₁₁H₁₀O₄
MW: 206.20 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00014375
CAS Number: 55620-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty