Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fast Blue BB base ≥95% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

A151391-1G A151391-25G
ALA151391-1GEA 9.41 USD InStock
ALA151391-1G ALA151391-25G
Fast Blue BB base ≥95% (by titration)
Fast Blue BB base


Formula: C₁₇H₂₀N₂O₃
Melting Pt: 101°C
MDL Number: MFCD00009091
CAS Number: 120-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty