Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-α,α,α-trifluoro-m-toluic acid ≥97% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

A151469-1G A151469-5G
ALA151469-1GEA 36.01 USD InStock
ALA151469-1G ALA151469-5G
5-Amino-α,α,α-trifluoro-m-toluic acid ≥97% (by HPLC, titration analysis)
5-Amino-α,α,α-trifluoro-m-toluic acid


Formula: C₈H₆F₃NO₂
MW: 205.14
Melting Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00236641
CAS Number: 328-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty