Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Amino-2-methylpyridinium iodide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151482-1G A151482-250MG A151482-5G
ALA151482-1GEA 78.76 USD InStock
ALA151482-1G ALA151482250MG ALA151482-5G
1-Amino-2-methylpyridinium iodide ≥98%
1-Amino-2-methylpyridinium iodide


Formula: C₆H₉IN₂
MW: 236.06 g/mol
Melting Pt: 152°C
Storage Temperature: Ambient
MDL Number: MFCD00955862
CAS Number: 7583-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty