Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diphenyl-1H-pyrazol-5-amine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151517-1G A151517-200MG A151517-250MG A151517-5G
ALA151517-1GEA 24.61 USD InStock
ALA151517-1G ALA151517200MG ALA151517250MG ALA151517-5G
1,3-Diphenyl-1H-pyrazol-5-amine ≥97%
1,3-Diphenyl-1H-pyrazol-5-amine


Formula: C₁₅H₁₃N₃
MW: 235.29 g/mol
Melting Pt: 128-132°C
MDL Number: MFCD00084878
CAS Number: 5356-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty