Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(alpha-Methylbenzyl)resorcinol ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151539-1G A151539-200MG A151539-25G A151539-5G
ALA151539-1GEA 46.46 USD InStock
ALA151539-1G ALA151539200MG ALA151539-25G ALA151539-5G
4-(alpha-Methylbenzyl)resorcinol ≥98%
4-(alpha-Methylbenzyl)resorcinol


Formula: C₁₄H₁₄O₂
Boiling Pt: 244°C
Melting Pt: 81 °C
MDL Number: MFCD21648466
CAS Number: 85-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty