Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-9H-pyrido[3,4-b]indole ≥97%

Supplier: ALADDIN SCIENTIFIC

A151563-100MG A151563-1G
ALA151563100MGEA 204.16 USD InStock
ALA151563100MG ALA151563-1G
3-Amino-9H-pyrido[3,4-b]indole ≥97%
3-Amino-9H-pyrido[3,4-b]indole


Formula: C₁₁H₉N₃
Melting Pt: 291°C
MDL Number: MFCD00209818
CAS Number: 73834-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty