Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-nitropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A151578-25G A151578-5G A151578-100G A151578-1G
ALA151578-25GEA 167.11 USD InStock
ALA151578-25G ALA151578-5G ALA151578-100G ALA151578-1G
2-Amino-5-bromo-3-nitropyridine ≥97%
2-Amino-5-bromo-3-nitropyridine


Formula: C₅H₄BrN₃O₂
Melting Pt: 209 °C
MDL Number: MFCD00047441
CAS Number: 6945-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty