Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-1-naphthol-3-sulfonic acid hydrate ≥95% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151598-100G A151598-25G
ALA151598-100GEA 97.76 USD InStock
ALA151598-100G ALA151598-25G
6-Amino-1-naphthol-3-sulfonic acid hydrate ≥95% (by HPLC)
6-Amino-1-naphthol-3-sulfonic acid hydrate


Formula: C₁₀H₉NO₄S
MW: 239.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003970
CAS Number: 87-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty