Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Anilino-4-hydroxy-2-naphthalenesulfonic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151600-25G A151600-500G A151600-5G
ALA151600-25GEA 49.31 USD InStock
ALA151600-25G ALA151600-500G ALA151600-5G
7-Anilino-4-hydroxy-2-naphthalenesulfonic acid ≥95%
7-Anilino-4-hydroxy-2-naphthalenesulfonic acid


Formula: C₁₆H₁₃NO₄S
MDL Number: MFCD00035714
CAS Number: 119-40-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty