Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1-propanol hydrochloride ≥95%

Supplier: ALADDIN SCIENTIFIC

A151629-100G A151629-25G A151629-5G
ALA151629-100GEA 117.71 USD InStock
ALA151629-100G ALA151629-25G ALA151629-5G
3-Amino-1-propanol hydrochloride ≥95%
3-Amino-1-propanol hydrochloride


Formula: C₃H₉NO
Storage Temperature: Ambient
MDL Number: MFCD00060203
CAS Number: 14302-46-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty