Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Aminobenzyl)phosphonic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151643-1G A151643-250MG
ALA151643-1GEA 271.61 USD InStock
ALA151643-1G ALA151643250MG
(4-Aminobenzyl)phosphonic acid ≥98%
(4-Aminobenzyl)phosphonic acid


Formula: C₇H₁₀NO₃P
Melting Pt: 325°C
Storage Temperature: Freezer
MDL Number: MFCD00070204
CAS Number: 5424-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty