Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Amino-2-benzoxazolinone ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A151644-100MG A151644-1G A151644-250MG A151644-5G
ALA151644100MGEA 21.76 USD InStock
ALA151644100MG ALA151644-1G ALA151644250MG ALA151644-5G
7-Amino-2-benzoxazolinone ≥95%
7-Amino-2-benzoxazolinone


Formula: C₇H₆N₂O₂
MW: 150.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09753616
CAS Number: 81282-60-2
EINECS: 462-910-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty