Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Angelic anhydride ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A151650-1G A151650-250MG A151650-5G
ALA151650-1GEA 91.11 USD InStock
ALA151650-1G ALA151650250MG ALA151650-5G
Angelic anhydride ≥98%
Angelic anhydride


Formula: C₁₀H₁₄O₃
Boiling Pt: 108°C
Density: 1.02 g/cm³
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00198080
CAS Number: 94487-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty