Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminopyridine-N-oxide ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

A151660-100MG A151660-1G
ALA151660100MGEA 27.46 USD InStock
ALA151660100MG ALA151660-1G
2-Aminopyridine-N-oxide ≥98% (by GC, titration analysis)
2-Aminopyridine N-oxide


Formula: C₅H₆N₂O
MW: 110.1139 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00059168
CAS Number: 14150-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty