Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetoxime benzoate ≥98%

Supplier: ALADDIN SCIENTIFIC

A151672-25G A151672-5G
ALA151672-25GEA 131.01 USD InStock
ALA151672-25G ALA151672-5G
Acetoxime benzoate ≥98%
Acetoxime benzoate


Formula: C₁₀H₁₁NO₂
Boiling Pt: 120°C
Melting Pt: 42 °C
MDL Number: MFCD00191697
CAS Number: 942-89-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty