Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-6-bromo-2-methylpyridine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151749-100G A151749-1G A151749-250MG A151749-25G A151749-5G
ALA151749-100GEA 929.01 USD InStock
ALA151749-100G ALA151749-1G ALA151749250MG ALA151749-25G ALA151749-5G
3-Amino-6-bromo-2-methylpyridine ≥97% (by GC)
3-Amino-6-bromo-2-methylpyridine


Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 108.0 ~112.0 °C
MDL Number: MFCD03095086
CAS Number: 126325-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty