Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentyl benzoate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

A151784-100ML A151784-25ML
ALA151784100MLEA 28.41 USD InStock
ALA151784100ML ALA151784-25ML
Pentyl benzoate ≥98% (by GC)
Pentyl benzoate


Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 260°C
Flash Pt: 130°C
MDL Number: MFCD00048843
CAS Number: 2049-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty