Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetamidopyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151789-1G A151789-250MG A151789-25G A151789-5G
ALA151789-1GEA 22.71 USD InStock
ALA151789-1G ALA151789250MG ALA151789-25G ALA151789-5G
3-Acetamidopyridine ≥98%
3-Acetamidopyridine


Formula: C₇H₈N₂O
Boiling Pt: 327°C
Melting Pt: 136 °C
CAS Number: 5867-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty