Acrylate PEG
Supplier: ALADDIN SCIENTIFIC
A163156-100MG A163156-500MG
ALA163156100MGEA 196.56 USD InStock
ALA163156100MG ALA163156500MG
Acrylate PEG
Acrylate PEG
Acetylthio PEG amine, also called thiolacetate PEG amine, is an amino PEG with protected thiol groups. Acetyl protected thiol group is stable in neutral condition and acetyl group can be deprotected from thiol group under acidic or alkaline conditions. Free thiol group is released upon deprotection of acetylthio. With thiol group protected, remaining functional groups, such as amines, azides, hydrazides, NHS, maleimides, DBCO, etc. can be used to react with other groups. PEGylated thioacetate derivatives are water soluble and can be used directly in aqueous buffer.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty