Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A165288-1G A165288-250MG A165288-5G
ALA165288-1GEA 26.51 USD InStock
ALA165288-1G ALA165288250MG ALA165288-5G
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate ≥98%
Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate is a heterocyclic building block.Ethyl 5-amino-1-phenyl-4-pyrazolecarboxylate may be used as internal standard for the GC-MS determination of etomidate [ethyl-1-(1-phenylethyl)-1H-imidazole-5-carboxylate] in mouse brain tissue.


Formula: C₇H₁₁N₃O₂
Melting Pt: 98-100°C
MDL Number: MFCD00020731
CAS Number: 16078-71-0
EINECS: 240-224-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty