Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylanthracene ≥98%

Supplier: ALADDIN SCIENTIFIC

A165488-1G A165488-250MG A165488-5G
ALA165488-1GEA 77.81 USD InStock
ALA165488-1G ALA165488250MG ALA165488-5G
2-Acetylanthracene ≥98%
2-Acetylanthracene
Product Application:2-Acetylanthracene is used as an organic fluorophore. It is also used as an intermediate in pharmaceuticals.


Formula: C₁₆H₁₂O
Melting Pt: 189-192 °C
Storage Temperature: Ambient
MDL Number: MFCD00156727
CAS Number: 10210-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty