Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azido-PEG12-acid ≥95%

Supplier: ALADDIN SCIENTIFIC

A166185-100MG A166185-25MG A166185-500MG
ALA166185100MGEA 242.16 USD InStock
ALA166185100MG ALA166185-25MG ALA166185500MG
Azido-PEG12-acid ≥95%
Azido-PEG12-acid


Formula: C₂₇H₅₃N₃O₁₄
MW: 643.721 g/mol
Storage Temperature: Freezer
MDL Number: MFCD21363301
CAS Number: 2152679-71-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty