Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-bromopyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A167068-1G A167068-25G A167068-5G
ALA167068-1GEA 13.21 USD InStock
ALA167068-1G ALA167068-25G ALA167068-5G
2-Amino-3-bromopyridine ≥97%
2-Amino-3-bromopyridine
Application:2-Amino-3-bromopyridine may be used to synthesize:2-acylamido-3-bromopyridines;2-anilino-3-bromopyridine;3-[(2-methoxyphenyl)ethynyl]pyridin-2-amine;3-[(4-methoxyphenyl)ethynyl]pyridin-2-amine;N-(bromopyridyl)amidines;carbolines via palladium catalyzed arylation followed by palladium catalyzed amination reaction;2-amino-3-cyanopyridine via palladium catalyzed cyanation reaction with potassium ferro-cyanide in the presence of 1,8-diazabicyclo[5.4.0]undec-7-ene [DBU];nitro-substituted N,N′-dipyridinylamines via palladium catalyzed coupling reaction with 2-chloro-3-nitropyridine in the presence of Xantphos ligand;2-amino-3-iodopyridine via reaction with sodium iodide in the presence of copper(I)iodide and trans-N,N′-dimethylcyclohexane-1,2-diamine;


Formula: C₅H₅BrN₂
MW: 173.01
Melting Pt: 63-67°C
Storage Temperature: Ambient
MDL Number: MFCD03095187
CAS Number: 13534-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty