Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetamido-2-deoxy-D-glucono-1,5-lactone ≥90% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A168205-25MG A168205-5MG A168205-100MG A168205-1MG
ALA168205-25MGEA 426.46 USD InStock
ALA168205-25MG ALA168205-5MG ALA168205100MG ALA168205-1MG
2-Acetamido-2-deoxy-D-glucono-1,5-lactone ≥90% (by HPLC)
2-Acetamido-2-deoxy-D-glucono-1,5-lactone
2-Acetamido-2-deoxy-D-glucono-1,5-lactone is a diagnostic agent that inhibits the activities of enzymes such as protein synthesis and cell division. It can be used to identify viral infections in animals, plants and marine microorganisms. 2-Acetamido-2-deoxy-D-glucono-1,5-lactone has been shown to inhibit the biochemical activity of enzymes in cells grown in culture. 2AADG is also a diagnostic agent that can be used to detect tumors in subcutaneous tissues due to its ability to inhibit the production of proteins essential for cell division.


Formula: C₈H₁₃NO₆
Storage Temperature: Freezer
MDL Number: MFCD00076154
CAS Number: 19026-22-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty