Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A169285-100MG A169285-1G A169285-250MG
ALA169285100MGEA 9.41 USD InStock
ALA169285100MG ALA169285-1G ALA169285250MG
tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate ≥97%
tert-Butyl-4-amino-4-carbamoylpiperidine-1-carboxylate


Formula: C₁₁H₂₁N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD04114477
CAS Number: 288154-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty