Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-trifluoromethyl-1,3-thiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A169774-250MG A169774-25G A169774-5G
ALA169774250MGEA 15.11 USD InStock
ALA169774250MG ALA169774-25G ALA169774-5G
2-Amino-4-trifluoromethyl-1,3-thiazole ≥97%
2-Amino-4-trifluoromethyl-1,3-thiazole


Formula: C₄H₃F₃N₂S
MW: 168.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00832759
CAS Number: 349-49-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty