Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopicolinic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A174297-5G A174297-1G A174297-25G
ALA174297-5GEA 47.41 USD InStock
ALA174297-5G ALA174297-1G ALA174297-25G
3-Aminopicolinic acid ≥97%
3-Aminopicolinic acid
Product Application:Employed as an important intermediate for syntheses material and pharmaceutical.


Formula: C₆H₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00090153
CAS Number: 1462-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty