Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-fluoroisonicotinic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A174940-100MG A174940-1G A174940-250MG
ALA174940100MGEA 40.76 USD InStock
ALA174940100MG ALA174940-1G ALA174940250MG
5-Amino-2-fluoroisonicotinic acid ≥97%
5-Amino-2-fluoroisonicotinic acid


Formula: C₆H₅FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03411724
CAS Number: 171178-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty