Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-1-methyl-1H-pyrazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A180010-100G A180010-1G A180010-25G A180010-5G
ALA180010-100GEA 559.46 USD InStock
ALA180010-100G ALA180010-1G ALA180010-25G ALA180010-5G
5-Amino-1-methyl-1H-pyrazole ≥98%
5-Amino-1-methyl-1H-pyrazole


Formula: C₄H₇N₃
MW: 97.12
Boiling Pt: 105 °C
Melting Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00068156
CAS Number: 1192-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty