Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-bromo-1-methyl-1H-pyrazole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A181564-1G A181564-250MG A181564-5G
ALA181564-1GEA 87.31 USD InStock
ALA181564-1G ALA181564250MG ALA181564-5G
3-Amino-4-bromo-1-methyl-1H-pyrazole ≥98%
3-Amino-4-bromo-1-methyl-1H-pyrazole


Formula: C₄H₆BrN₃
Melting Pt: 96°C
MDL Number: MFCD00663065
CAS Number: 146941-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty