Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dimethoxy-1,3,5-triazine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A181911-250MG A181911-25G
ALA181911250MGEA 9.41 USD InStock
ALA181911250MG ALA181911-25G
2-Amino-4,6-dimethoxy-1,3,5-triazine ≥98%
2-Amino-4,6-dimethoxy-1,3,5-triazine


Formula: C₅H₈N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00052765
CAS Number: 16370-63-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty