Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylphenylboronic acid pinacol ester ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

A182048-25G A182048-5G A182048-1G
ALA182048-25GEA 337.16 USD InStock
ALA182048-25G ALA182048-5G ALA182048-1G
4-Acetylphenylboronic acid pinacol ester ≥96%
4-Acetylphenylboronic acid pinacol ester


Formula: C₁₄H₁₉BO₃
MDL Number: MFCD05863922
CAS Number: 171364-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty